Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich tert-Dodecylmercaptan

Catalog No. 4715852L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
2 L
18 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-158-52L 2 L
47-158-5100ML 100 mL
47-158-518L 18 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 47-158-52L Supplier Sigma Aldrich Supplier No. 4715852L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Product of Arkema Inc.

Specifications

CAS 25103-58-6
Density 0.86 g/mL (at 20°C)
Boiling Point 227°C to 248°C
Molecular Formula C12H26S
Linear Formula C12H25SH
Quantity 2 L
Synonym tert-Dodecanethiol
Molecular Weight (g/mol) 202.4
Percent Purity 98.5%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.