Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Octyl Isothiocyanate 94%, Thermo Scientific™

Catalog No. AC129511000
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC129511000 100 g
AC129510250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC129511000 Supplier Thermo Scientific Chemicals Supplier No. 129511000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17701-76-7
Molecular Formula C9H17NS
Molecular Weight (g/mol) 171.302
InChI Key KUGHMFIBHMZICO-UHFFFAOYSA-N
Synonym tert-octyl isothiocyanate, tert-octylisothiocyanate, 1,1,3,3-tetramethylbutyl isothiocyanate, pentane,2-isothiocyanato-2,4,4-trimethyl, pentane, 2-isothiocyanato-2,4,4-trimethyl, t-octylisothiocyanate, 2-isothiocyanato-2,4,4-trimethyl-pentane
PubChem CID 87255
IUPAC Name 2-isothiocyanato-2,4,4-trimethylpentane
SMILES CC(C)(C)CC(C)(C)N=C=S

Specifications

Density 1.199g/mL
Infrared Spectrum Authentic
Assay Percent Range 94%
Refractive Index 1.4830 to 1.4880 (20°C, 589nm)
Linear Formula (CH3)3CCH2C(CH3)2NCS
Quantity 100 g
Formula Weight 171.3
Percent Purity ≥92%
Physical Form Liquid
Chemical Name or Material tert-Octyl isothiocyanate, 94%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 241-710-2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.