Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Tetradecanethiol (mixture of isomers) 97.0+%, TCI America™
SDP

Catalog No. T008325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T008325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T008325G Supplier TCI America Supplier No. T008325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 28983-37-1
Molecular Formula C14H30S
Molecular Weight (g/mol) 230.454
MDL Number MFCD00041145
InChI Key LMDDHLWHSDZGIH-UHFFFAOYSA-N
Synonym tert-Tetradecyl Mercaptan
PubChem CID 21493711
IUPAC Name 2-methyltridecane-2-thiol
SMILES CCCCCCCCCCCC(C)(C)S

Specifications

Color Yellow
Quantity 25 g
Formula Weight 230.45
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material tert-Tetradecanethiol (mixture of isomers)
EINECSNumber (2)-0464
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.