Learn More
Medchemexpress LLC Tertiapin-Q | 910044-56-3 | 99.8% | 2452.00 | 1 MG
SDP
Catalog No. 5000423774
Encompass_Preferred
Tertiapin-Q is a highly selective blocker of G protein-coupled inwardly rectifying potassium (GIRK1/4) heterodimer and renal outer medullary potassium channel (ROMK1/Kir1.1). It also potently and selectively blocks Kir3.1-Kir3.4 channels and calcium activated large conductance potassium channels. Tertiapin-Q significantly prolongs the atrial effective refractory period in control and rapid atrial pacing rabbits, with a greater effect in the latter. Additionally, it acts as a muscarinic acetylcholine receptor-operated K+ current (IK,Ach) blocker.
  • Highly selective blocker of GIRK1/4 heterodimer and ROMK1 (Kir1.1)
  • Potently blocks Kir3.1-Kir3.4 channels and calcium activated large conductance potassium channels
  • Prolongs atrial effective refractory period
  • Acts as a muscarinic acetylcholine receptor-operated K+ current blocker
Catalog No. 50-004-23774 Supplier Medchemexpress LLC Supplier No. HYP12751MG
Tertiapin-Q is a highly selective blocker of G protein-coupled inwardly rectifying potassium (GIRK1/4) heterodimer and renal outer medullary potassium channel (ROMK1/Kir1.1). It also potently and selectively blocks Kir3.1-Kir3.4 channels and calcium activated large conductance potassium channels. Tertiapin-Q significantly prolongs the atrial effective refractory period in control and rapid atrial pacing rabbits, with a greater effect in the latter. Additionally, it acts as a muscarinic acetylcholine receptor-operated K+ current (IK,Ach) blocker.
  • Highly selective blocker of GIRK1/4 heterodimer and ROMK1 (Kir1.1)
  • Potently blocks Kir3.1-Kir3.4 channels and calcium activated large conductance potassium channels
  • Prolongs atrial effective refractory period
  • Acts as a muscarinic acetylcholine receptor-operated K+ current blocker