Learn More
Abcam Tertiapin Q, blocker of inward-rectifier r K+ channels, 1MG
Catalog No. 502707592
Encompass_Preferred
MW 2295.8 Da, Purity >95%. Tertiapin Q, blocker of inward-rectifier r K+ channels. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7592 Supplier Abcam Supplier No. AB1204321MG
MW 2295.8 Da, Purity >95%. Tertiapin Q, blocker of inward-rectifier r K+ channels. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement