Learn More
Selleck Chemical LLC Tertiapin-Q-E0016-25MG
SDP
Catalog No. 503135613
Encompass_Preferred
Tertiapin-Q (TPNQ) is a derivative of tertiapin and inhibits BK-type K channels in a use- and concentration-dependent manner Tertiapin-Q also binds to ROMK1 (Kir1 1) and GIRK1/4 (Kir3 1/3 4) channels with Ki values of 1 3 nM and 13 3 nM respectively
Catalog No. 50-313-5613 Supplier Selleck Chemical LLC Supplier No. E001625MG
Tertiapin-Q (TPNQ) is a derivative of tertiapin and inhibits BK-type K channels in a use- and concentration-dependent manner Tertiapin-Q also binds to ROMK1 (Kir1 1) and GIRK1/4 (Kir3 1/3 4) channels with Ki values of 1 3 nM and 13 3 nM respectively