Learn More
Medchemexpress LLC HY-112619 10mg Medchemexpress, TES-991 CAS:1883602-20-7 Purity:>98%
SDP
Catalog No. 501932428
Encompass_Preferred
Medchemexpress, HY-112619 10mg TES-991 CAS:1883602-20-7 TES-991 is a potent and selective human α‑Amino-β-carboxymuconate-ε-semialdehyde Decarboxylase (ACMSD) inhibitor, with an IC 50 of 3 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2428 Supplier Medchemexpress LLC Supplier No. HY11261910MG
Medchemexpress, HY-112619 10mg TES-991 CAS:1883602-20-7 TES-991 is a potent and selective human α‑Amino-β-carboxymuconate-ε-semialdehyde Decarboxylase (ACMSD) inhibitor, with an IC 50 of 3 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.