Learn More
Medchemexpress LLC TES-991 | 1883602-20-7 | 99.3% | 393.45 | 1 ML
SDP
Catalog No. 5000436055
Encompass_Preferred
TES-991 is a potent and selective human α Amino-β-carboxymuconate-ε-semialdehyde Decarboxylase (ACMSD) inhibitor, with an IC50 of 3 nM. It is able to significantly increase intracellular NAD+ levels.
  • Potent and selective human α Amino-β-carboxymuconate-ε-semialdehyde Decarboxylase (ACMSD) inhibitor with an IC50 of 3 nM.
  • Significantly increases intracellular NAD+ levels.
  • Low blood clearance after intravenous administration.
  • Moderate systemic exposure with the 2H-tetrazole analogue.
Catalog No. 50-004-36055 Supplier Medchemexpress LLC Supplier No. HY11261910MM/1ML
TES-991 is a potent and selective human α Amino-β-carboxymuconate-ε-semialdehyde Decarboxylase (ACMSD) inhibitor, with an IC50 of 3 nM. It is able to significantly increase intracellular NAD+ levels.
  • Potent and selective human α Amino-β-carboxymuconate-ε-semialdehyde Decarboxylase (ACMSD) inhibitor with an IC50 of 3 nM.
  • Significantly increases intracellular NAD+ levels.
  • Low blood clearance after intravenous administration.
  • Moderate systemic exposure with the 2H-tetrazole analogue.