Learn More
Medchemexpress LLC TES-991 | 1883602-20-7 | 99.7% | 393.45 | 25 MG
SDP
Catalog No. 5000439783
Encompass_Preferred
TES-991 is a potent and selective human α Amino-β-carboxymuconate-ε-semialdehyde Decarboxylase (ACMSD) inhibitor, with an IC50 of 3 nM.
  • Potent and selective human α Amino-β-carboxymuconate-ε-semialdehyde Decarboxylase (ACMSD) inhibitor with an IC50 of 3 nM.
  • Significantly increases intracellular NAD+ levels.
  • Shows low blood clearance with low volumes of distribution and half-lives of approximately 4.0 and 5.0 hours after intravenous administration (0.5 mg/kg).
  • Blood concentrations are quantifiable for up to 8 hours after oral administration (5 mg/kg).
  • Exhibits moderate systemic exposure for the 2H-tetrazole analogue and good systemic exposure for the free acid.
Catalog No. 50-004-39783 Supplier Medchemexpress LLC Supplier No. 25MGHY112619
TES-991 is a potent and selective human α Amino-β-carboxymuconate-ε-semialdehyde Decarboxylase (ACMSD) inhibitor, with an IC50 of 3 nM.
  • Potent and selective human α Amino-β-carboxymuconate-ε-semialdehyde Decarboxylase (ACMSD) inhibitor with an IC50 of 3 nM.
  • Significantly increases intracellular NAD+ levels.
  • Shows low blood clearance with low volumes of distribution and half-lives of approximately 4.0 and 5.0 hours after intravenous administration (0.5 mg/kg).
  • Blood concentrations are quantifiable for up to 8 hours after oral administration (5 mg/kg).
  • Exhibits moderate systemic exposure for the 2H-tetrazole analogue and good systemic exposure for the free acid.