Learn More
Medchemexpress LLC TES-991 | 1883602-20-7 | 99.7% | 393.45 | 50 MG
SDP
Catalog No. 5000437654
Encompass_Preferred
TES-991 is a potent and selective human α-Amino-β-carboxymuconate-ε-semialdehyde Decarboxylase (ACMSD) inhibitor, with an IC50 of 3 nM. It is for research use only. This compound significantly increases intracellular NAD+ levels, and in vivo studies show low blood clearance and moderate systemic exposure after oral administration.
  • Potent and selective ACMSD inhibitor (IC50 of 3 nM).
  • Significantly increases intracellular NAD+ levels.
  • Low blood clearance and volumes of distribution (in vivo studies).
  • Moderate systemic exposure after oral administration (in vivo studies).
  • Stable for 3 years at -20°C as powder.
  • Stable for 2 years at -80°C in solvent.
Catalog No. 50-004-37654 Supplier Medchemexpress LLC Supplier No. HY11261950MG
TES-991 is a potent and selective human α-Amino-β-carboxymuconate-ε-semialdehyde Decarboxylase (ACMSD) inhibitor, with an IC50 of 3 nM. It is for research use only. This compound significantly increases intracellular NAD+ levels, and in vivo studies show low blood clearance and moderate systemic exposure after oral administration.
  • Potent and selective ACMSD inhibitor (IC50 of 3 nM).
  • Significantly increases intracellular NAD+ levels.
  • Low blood clearance and volumes of distribution (in vivo studies).
  • Moderate systemic exposure after oral administration (in vivo studies).
  • Stable for 3 years at -20°C as powder.
  • Stable for 2 years at -80°C in solvent.