Learn More
Alkali Scientific TES, Free Acid [2 Tris(hydroxymethyl) methyl 2 amino 1 ethanesulfonic acid], 100 G
SDP
Catalog No. 5000660474
Encompass_Preferred
TES, or Tris(hydroxymethyl) methyl 2-amino-1-ethanesulfonic acid, is available in free acid form for use in laboratory buffer preparations. The 100g size is ideal for labs that require frequent buffer preparation for a variety of biochemical assays. TES is commonly used in applications where a stable pH is required, including protein studies, enzyme assays, and chromatography. The high purity of TES ensures that it will not interfere with sensitive reactions, allowing researchers to obtain reproducible and accurate results. This buffering agent is particularly useful for applications in which minimal UV absorbance is essential for measurements, such as in spectrophotometric assays.
Catalog No. 50-006-60474 Supplier Alkali Scientific Supplier No. TES004CP
TES, or Tris(hydroxymethyl) methyl 2-amino-1-ethanesulfonic acid, is available in free acid form for use in laboratory buffer preparations. The 100g size is ideal for labs that require frequent buffer preparation for a variety of biochemical assays. TES is commonly used in applications where a stable pH is required, including protein studies, enzyme assays, and chromatography. The high purity of TES ensures that it will not interfere with sensitive reactions, allowing researchers to obtain reproducible and accurate results. This buffering agent is particularly useful for applications in which minimal UV absorbance is essential for measurements, such as in spectrophotometric assays.