Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TES, Free Acid, G-Biosciences
SCI_ED

Catalog No. 01001058
Change view
Click to view available options
Quantity:
500 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-058 500 g
01-001-082 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-001-058 Supplier G-Biosciences Supplier No. RC826
Add to Cart
Add to Cart

TES has been used in TES buffer to determine anti-collagenase activity of Marula stems.

It has also been used in buffers for cell culture medium.

Chemical Identifiers

Molecular Weight (g/mol) 229.25 g/mol

Specifications

Quantity 500 g
Form Powder
Percent Purity >95%
Grade Reagent
Chemical Name or Material TES
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.