Learn More
Medchemexpress LLC TES sodium | 70331-82-7 | 98.0% | 251.23 | 5 G
SDP
Catalog No. 5000401974
Encompass_Preferred
TES sodium is an excellent hydrogen ion buffer. It possesses characteristics such as an appropriate dissociation constant, a low metal-binding constant, and high water solubility. TES sodium is suitable for research involving mammalian cell culture, viruses, and similar applications.
  • Excellent hydrogen ion buffer.
  • Appropriate dissociation constant.
  • Low metal-binding constant.
  • High water solubility.
  • Can be used in research of mammalian cell culture and viruses.
  • Shows similar or even more effective results than NaHCO3-CO2-buffered medium in terms of colony-forming efficiency, colony size, and cell morphology for several mammalian cell lines (HeLa S3, KB, BHK 21-Cl3, 3Y1-B Cl-1).
  • Demonstrates similar efficiency to NaHCO3-CO2-buffered medium in SV40 virus plaque assay with monkey kidney CV-1 cells, with shorter required time.
Catalog No. 50-004-01974 Supplier Medchemexpress LLC Supplier No. HY23430A5G
TES sodium is an excellent hydrogen ion buffer. It possesses characteristics such as an appropriate dissociation constant, a low metal-binding constant, and high water solubility. TES sodium is suitable for research involving mammalian cell culture, viruses, and similar applications.
  • Excellent hydrogen ion buffer.
  • Appropriate dissociation constant.
  • Low metal-binding constant.
  • High water solubility.
  • Can be used in research of mammalian cell culture and viruses.
  • Shows similar or even more effective results than NaHCO3-CO2-buffered medium in terms of colony-forming efficiency, colony size, and cell morphology for several mammalian cell lines (HeLa S3, KB, BHK 21-Cl3, 3Y1-B Cl-1).
  • Demonstrates similar efficiency to NaHCO3-CO2-buffered medium in SV40 virus plaque assay with monkey kidney CV-1 cells, with shorter required time.