Learn More
Research Products International Corp TES, Sodium Salt, 25 G
SDP
Catalog No. 501978437
Encompass_Preferred
TES (N-[Tris(hydroxymethyl)methyl]-2-aminoethanesulfonic acid) Sodium Salt is a commonly used buffering agent in life science research. It helps to maintain a stable pH environment in biological experiments, such as cell culture, protein purification, and enzyme assays. 
TES Sodium Salt is advantageous due to its ability to resist changes in pH over a wide range of temperatures, making it suitable for various experimental conditions. 
Additionally, its low UV absorbance and minimal interference with biological molecules make it a preferred choice in many biochemical applications
Catalog No. 50-197-8437 Supplier Research Products International Corp Supplier No. T3102025.0
TES (N-[Tris(hydroxymethyl)methyl]-2-aminoethanesulfonic acid) Sodium Salt is a commonly used buffering agent in life science research. It helps to maintain a stable pH environment in biological experiments, such as cell culture, protein purification, and enzyme assays. 
TES Sodium Salt is advantageous due to its ability to resist changes in pH over a wide range of temperatures, making it suitable for various experimental conditions. 
Additionally, its low UV absorbance and minimal interference with biological molecules make it a preferred choice in many biochemical applications