Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TES (White Crystals), Fisher BioReagents

Catalog No. BP309100 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP309-100 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP309-100

missing translation for 'validMainframeMsg'

This biological buffer has a usable pH range of 6.8 to 8.2.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7365-44-8
Molecular Formula C6H15NO6S
Molecular Weight (g/mol) 229.247
InChI Key JOCBASBOOFNAJA-UHFFFAOYSA-N
Synonym N-Tris(hydroxymethyl)methyl-2-aminoethanesulfonic Acid
PubChem CID 81831
ChEBI CHEBI:44356
IUPAC Name 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]ethanesulfonic acid
SMILES C(CS(=O)(=O)O)NC(CO)(CO)CO

Specifications

Melting Point 223°C
Color White
Decomposition Information >220°C
pH 4.0 to 4.8
Physical Form Powder or Solid
Quantity 100 g
Assay Percent Range ≥98 %
Identification Pass Test
Packaging Glass Bottle
Merck Index 15, 9319
Vapor Pressure Negligible
Formula Weight 229.25
Percent Purity ≥98%
Chemical Name or Material TES
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Irritating to eyes, respiratory system and skin. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Obtain medical attention. . .

NFPA

Health:2
Flammability:1
Instability:0
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.