Learn More
MSC TESA Brown & Sharpe Combination Square Set, 2 Piece, 12 inch, Hardened steel
Catalog No. 5000532553
Encompass
This combination square set is designed for precise measurement and marking tasks. It features a hardened steel blade and square head, ensuring durability and accuracy for various applications.
  • Two pieces for versatile use.
  • 12 inch blade length for extended reach.
  • Hardened steel construction for durability and accuracy.
  • 4R graduation style for precise readings.
Catalog No. 50-005-32553 Supplier MSC Supplier No. 06505655
This combination square set is designed for precise measurement and marking tasks. It features a hardened steel blade and square head, ensuring durability and accuracy for various applications.
  • Two pieces for versatile use.
  • 12 inch blade length for extended reach.
  • Hardened steel construction for durability and accuracy.
  • 4R graduation style for precise readings.