Learn More
MSC TESA Brown & Sharpe Dial Caliper, 8 in, 0.001 in, Stainless Steel
Catalog No. 5000535020
Encompass
This dial caliper offers an 8 in maximum measurement with a 0.001 in graduation and 0.1 in per revolution. Constructed with stainless steel jaws, it is designed for performing depth, inside, outside, and step measurements.
  • Performs outside, step, inside, and depth measurements.
  • Constructed from stainless steel.
  • Integrated shockproof system.
  • Adheres to ISO 13385-1:2019 standards.
  • Features a silver dial face and gray dial case.
  • Jaw length of 1.9 in.
Catalog No. 50-005-35020 Supplier MSC Supplier No. 63739585
This dial caliper offers an 8 in maximum measurement with a 0.001 in graduation and 0.1 in per revolution. Constructed with stainless steel jaws, it is designed for performing depth, inside, outside, and step measurements.
  • Performs outside, step, inside, and depth measurements.
  • Constructed from stainless steel.
  • Integrated shockproof system.
  • Adheres to ISO 13385-1:2019 standards.
  • Features a silver dial face and gray dial case.
  • Jaw length of 1.9 in.