Learn More
MSC TESA Brown & Sharpe Dial Drop Indicator, 0.2000" Max Measurement, 0.0001" Graduation, 0-5-0 Dial Reading, 2-1/4" Dial Diameter
Catalog No. 5000512185
Encompass
This precision Dial Drop Indicator is engineered for accurate measurement applications. It features a robust design with a white dial, a central lug back, and a revolution counter, making it suitable for various laboratory and industrial settings. The AGD 2 series indicators are a popular choice, offering reliable performance with a ball contact tip and tolerance pointers.
  • Maximum measurement of 0.2000 inch
  • Graduation of 0.0001 inch
  • 0-5-0 dial reading
  • 2-1/4 inch dial diameter
  • White dial face
  • Features a central lug back
  • Equipped with a revolution counter
  • Includes tolerance pointers
  • Comes standard with a ball contact tip
  • Conforms to AGD Group 2 standards
Catalog No. 50-005-12185 Supplier MSC Supplier No. 67570077
This precision Dial Drop Indicator is engineered for accurate measurement applications. It features a robust design with a white dial, a central lug back, and a revolution counter, making it suitable for various laboratory and industrial settings. The AGD 2 series indicators are a popular choice, offering reliable performance with a ball contact tip and tolerance pointers.
  • Maximum measurement of 0.2000 inch
  • Graduation of 0.0001 inch
  • 0-5-0 dial reading
  • 2-1/4 inch dial diameter
  • White dial face
  • Features a central lug back
  • Equipped with a revolution counter
  • Includes tolerance pointers
  • Comes standard with a ball contact tip
  • Conforms to AGD Group 2 standards