Learn More
MSC TESA Brown & Sharpe Dial Indicator & Base Kit, Black Dial, 0.001" Graduation, Magnetic Base
Catalog No. 5000528234
Encompass
This TESA Brown & Sharpe Dial Indicator & Base Kit is designed for precise measurements in various applications. It features a black dial with 0.001" graduation, and includes a Mighty Mag magnetic base for secure positioning.
  • Dial indicator type
  • Magnetic base style
  • 0.001" graduation
  • 2-1/4" dial diameter
  • Black dial color
  • 0-100 dial reading
  • 4" base length
  • 1" base width
  • 1" base height
  • Includes Mighty Mag magnetic base set and indicator
Catalog No. 50-005-28234 Supplier MSC Supplier No. 53987426
This TESA Brown & Sharpe Dial Indicator & Base Kit is designed for precise measurements in various applications. It features a black dial with 0.001" graduation, and includes a Mighty Mag magnetic base for secure positioning.
  • Dial indicator type
  • Magnetic base style
  • 0.001" graduation
  • 2-1/4" dial diameter
  • Black dial color
  • 0-100 dial reading
  • 4" base length
  • 1" base width
  • 1" base height
  • Includes Mighty Mag magnetic base set and indicator