Learn More
MSC TESA Brown & Sharpe Dial Indicator Kit, 0.0300 in Max Measurement, 0.0005 in Graduation, Universal Holder
Catalog No. 5000516829
Encompass
This TESA Brown & Sharpe dial indicator and base kit is designed for precise measurement applications. It includes a BesTest dial indicator and a universal holder, making it suitable for a variety of setups and measurements in industrial settings.
  • Dial indicator for accurate readings.
  • Maximum measurement capacity of 0.0300 in.
  • Graduation of 0.000500 in for fine measurements.
  • Features a 0-15-0 dial reading for clear interpretation.
  • Includes a universal holder for versatile positioning.
  • Kit consists of 4 pieces for comprehensive measuring tasks.
Catalog No. 50-005-16829 Supplier MSC Supplier No. 06412662
This TESA Brown & Sharpe dial indicator and base kit is designed for precise measurement applications. It includes a BesTest dial indicator and a universal holder, making it suitable for a variety of setups and measurements in industrial settings.
  • Dial indicator for accurate readings.
  • Maximum measurement capacity of 0.0300 in.
  • Graduation of 0.000500 in for fine measurements.
  • Features a 0-15-0 dial reading for clear interpretation.
  • Includes a universal holder for versatile positioning.
  • Kit consists of 4 pieces for comprehensive measuring tasks.