Learn More
MSC TESA Brown & Sharpe Digital 3-Point Inside Micrometer, 10-12 mm Range, 0.001 mm Resolution, 4 μm Accuracy
Catalog No. 5000529981
Encompass
This digital 3-point inside micrometer is designed for precise internal measurements. It features a measuring range of 10 to 12 mm, a resolution of 0.001 mm, and an accuracy of 4 μm. This tool is suitable for laboratory and industrial applications.
  • Automatic shut down after 10 min
  • Display setting is retained as long as power supply remains stable
  • Opto-coupled RS-232 interface bidirectional
  • Metric/inch conversion
  • Floating zero
  • IP54 rated
  • Includes (1) 3V lithium battery and (1) plastic case
  • Hardened steel spindle material
  • Hardened steel anvil material
  • Standards: DIN 863 T4 (C1)
  • Curved contact point shape
  • LCD display type
  • Includes certificate of inspection and declaration of conformity
Catalog No. 50-005-29981 Supplier MSC Supplier No. 06226344
This digital 3-point inside micrometer is designed for precise internal measurements. It features a measuring range of 10 to 12 mm, a resolution of 0.001 mm, and an accuracy of 4 μm. This tool is suitable for laboratory and industrial applications.
  • Automatic shut down after 10 min
  • Display setting is retained as long as power supply remains stable
  • Opto-coupled RS-232 interface bidirectional
  • Metric/inch conversion
  • Floating zero
  • IP54 rated
  • Includes (1) 3V lithium battery and (1) plastic case
  • Hardened steel spindle material
  • Hardened steel anvil material
  • Standards: DIN 863 T4 (C1)
  • Curved contact point shape
  • LCD display type
  • Includes certificate of inspection and declaration of conformity