Learn More
MSC TESA Brown & Sharpe Double Square, 4 Inch Blade Length, 4R Graduation
Catalog No. 5000529963
Encompass
This double square is designed for precise measurements and marking in various applications. It features a durable construction and clear, easy-to-read graduations, making it a reliable tool for professionals.
  • 4 inch blade length
  • 4R graduation style
  • Graduations of 1/64, 1/32, 1/16, and 1/8 inch for detailed work
  • High quality and easy to read markings
  • Ideal for workshop and measurement tasks
Catalog No. 50-005-29963 Supplier MSC Supplier No. 00376160
This double square is designed for precise measurements and marking in various applications. It features a durable construction and clear, easy-to-read graduations, making it a reliable tool for professionals.
  • 4 inch blade length
  • 4R graduation style
  • Graduations of 1/64, 1/32, 1/16, and 1/8 inch for detailed work
  • High quality and easy to read markings
  • Ideal for workshop and measurement tasks