Learn More
MSC TESA Brown & Sharpe Edge Finder, 0.5 in Head, 1/2 in Shank, Mechanical
Catalog No. 5000529425
Encompass
This mechanical edge finder is designed for precise machine setup, featuring a 0.5 in head diameter and a 0.5 in shank diameter. It offers high accuracy for consistent results in various machining applications.
  • Mechanical operation for reliable performance.
  • 0.5 in head diameter for accurate contact.
  • 0.5 in shank diameter ensures compatibility with standard tool holders.
  • Provides 0.0005 in accuracy for precise measurements.
  • Equipped with a cylindrical contact type for efficient edge detection.
Catalog No. 50-005-29425 Supplier MSC Supplier No. 02035293
This mechanical edge finder is designed for precise machine setup, featuring a 0.5 in head diameter and a 0.5 in shank diameter. It offers high accuracy for consistent results in various machining applications.
  • Mechanical operation for reliable performance.
  • 0.5 in head diameter for accurate contact.
  • 0.5 in shank diameter ensures compatibility with standard tool holders.
  • Provides 0.0005 in accuracy for precise measurements.
  • Equipped with a cylindrical contact type for efficient edge detection.