Learn More
MSC TESA Brown & Sharpe Edge Finder, Mechanical, Audible, 10 mm Head Dia, 10 mm Shank Dia
Catalog No. 5000516492
Encompass
This edge finder is a mechanical device designed for precise location gaging on flat edges. It features a cylindrical head with a 10 mm diameter and a 10 mm shank diameter, providing accurate measurement for various applications.
  • Mechanical power type for reliable operation.
  • Cylindrical head contact for accurate edge location.
  • 10 mm head diameter.
  • 10 mm shank diameter.
  • Single edge end type.
  • Audible alert mechanism for user feedback.
  • Non-magnetic design.
Catalog No. 50-005-16492 Supplier MSC Supplier No. 02035285
This edge finder is a mechanical device designed for precise location gaging on flat edges. It features a cylindrical head with a 10 mm diameter and a 10 mm shank diameter, providing accurate measurement for various applications.
  • Mechanical power type for reliable operation.
  • Cylindrical head contact for accurate edge location.
  • 10 mm head diameter.
  • 10 mm shank diameter.
  • Single edge end type.
  • Audible alert mechanism for user feedback.
  • Non-magnetic design.