Learn More
MSC TESA Brown & Sharpe Electronic Caliper, 200 mm, 8 in, 0.01 mm Resolution, Stainless Steel
Catalog No. 5000527109
Encompass
This electronic caliper is constructed from stainless steel and is designed for precise measurements including depth, inside diameter, outside diameter, and step. It features an IP40 rating for protection against solid objects and offers a measurement range of 0-200 mm (0-8 in) with high resolution and accuracy.
  • Resolution of ±0.0005 in (±0.0100 mm)
  • Accuracy of ±0.0010 in (±0.02 mm)
  • Built-in wireless data output
  • IP40 rated
  • Stainless steel construction for caliper and jaws
  • Jaw length of 50.00 mm
  • Measures inside diameter, step, outside diameter, and depth
  • Knife-edge jaw type with thumb wheel adjustment
  • Powered by one CR2032 lithium battery
  • Includes declaration of conformity and measuring report
  • Meets ISO 13385-1 standards
Catalog No. 50-005-27109 Supplier MSC Supplier No. 11517836
This electronic caliper is constructed from stainless steel and is designed for precise measurements including depth, inside diameter, outside diameter, and step. It features an IP40 rating for protection against solid objects and offers a measurement range of 0-200 mm (0-8 in) with high resolution and accuracy.
  • Resolution of ±0.0005 in (±0.0100 mm)
  • Accuracy of ±0.0010 in (±0.02 mm)
  • Built-in wireless data output
  • IP40 rated
  • Stainless steel construction for caliper and jaws
  • Jaw length of 50.00 mm
  • Measures inside diameter, step, outside diameter, and depth
  • Knife-edge jaw type with thumb wheel adjustment
  • Powered by one CR2032 lithium battery
  • Includes declaration of conformity and measuring report
  • Meets ISO 13385-1 standards