Learn More
MSC TESA Brown & Sharpe Electronic Inside Micrometer, 0.3200-0.4000 Inch, 0.00005 Inch Resolution, IP40/IP54
Catalog No. 5000533263
Encompass
TESA Brown & Sharpe Electronic Inside Micrometer provides precise internal measurements with a range of 0.3200 to 0.4000 inches (8.00 to 10.00 mm). It offers high resolution and accuracy, making it suitable for demanding applications. The device also features data output capabilities and comes with a protective plastic case.
  • Measures internal dimensions with high precision.
  • Features a measurement range from 0.3200 to 0.4000 inches (8.00 to 10.00 mm).
  • Offers a resolution of 0.00005 inches.
  • Ensures accuracy of 0.00016 inches (4 micrometers).
  • Equipped with 3 contact points for stable readings.
  • Provides data output functionality.
  • Includes a plastic case for protection and storage.
  • Operates on a CR2032 battery.
  • Features an Ingress Protection rating of IP40/IP54.
Catalog No. 50-005-33263 Supplier MSC Supplier No. 06226336
TESA Brown & Sharpe Electronic Inside Micrometer provides precise internal measurements with a range of 0.3200 to 0.4000 inches (8.00 to 10.00 mm). It offers high resolution and accuracy, making it suitable for demanding applications. The device also features data output capabilities and comes with a protective plastic case.
  • Measures internal dimensions with high precision.
  • Features a measurement range from 0.3200 to 0.4000 inches (8.00 to 10.00 mm).
  • Offers a resolution of 0.00005 inches.
  • Ensures accuracy of 0.00016 inches (4 micrometers).
  • Equipped with 3 contact points for stable readings.
  • Provides data output functionality.
  • Includes a plastic case for protection and storage.
  • Operates on a CR2032 battery.
  • Features an Ingress Protection rating of IP40/IP54.