Learn More
MSC TESA Brown & Sharpe Horizontal Dial Test Indicator, 0.008 in, 0.0001 in Graduation
Catalog No. 5000529427
Encompass
This horizontal dial test indicator is a precision measuring instrument for accurate comparative measurements. It features an integrated shockproof system and low measuring force, making it suitable for demanding industrial applications. The set includes carbide contact points, dovetail extensions, and a certificate of accuracy.
  • Integrated shockproof system.
  • Low measuring force.
  • Three carbide contact points (0.040 in, 0.080 in, 0.120 in).
  • Two dovetail extensions and contact point wrench.
  • Certificate of accuracy and traceability.
  • Durable molded case for protection and storage.
  • Conforms to DIN 2270 and EN ISO 9493 standards.
  • Horizontal indicator style.
  • Lever type movement.
  • Rotating dial.
  • Analogue display.
Catalog No. 50-005-29427 Supplier MSC Supplier No. 06433494
This horizontal dial test indicator is a precision measuring instrument for accurate comparative measurements. It features an integrated shockproof system and low measuring force, making it suitable for demanding industrial applications. The set includes carbide contact points, dovetail extensions, and a certificate of accuracy.
  • Integrated shockproof system.
  • Low measuring force.
  • Three carbide contact points (0.040 in, 0.080 in, 0.120 in).
  • Two dovetail extensions and contact point wrench.
  • Certificate of accuracy and traceability.
  • Durable molded case for protection and storage.
  • Conforms to DIN 2270 and EN ISO 9493 standards.
  • Horizontal indicator style.
  • Lever type movement.
  • Rotating dial.
  • Analogue display.