Learn More
MSC TESA Brown & Sharpe Horizontal Dial Test Indicator, 0.0300 inch Max, 0-15-0 Dial, 0.001 inch Graduation
Catalog No. 5000532591
Encompass
This horizontal dial test indicator from the BesTest series offers precise measurements with a maximum range of 0.0300 inch, a 0-15-0 dial reading, and a 0.001 inch graduation. It is designed for laboratory and industrial applications, adhering to DIN 2270 and EN ISO 9493 standards.
  • Horizontal indicator style
  • 1 inch dial diameter
  • Black dial color
  • Lever type with integrated shockproof system
  • Analogue display
  • Low measuring force
  • Rotating dial
  • Includes one contact point
  • 1/2 inch contact point length
  • 0.0800 inch ball diameter
  • 0.0300 inch range per revolution
  • Complies with DIN 2270 and EN ISO 9493 standards
  • Comes with a measurement report, wrench, mounting rod, declaration of conformity certificate, and a 0.08 inch insert
Catalog No. 50-005-32591 Supplier MSC Supplier No. 09989740
This horizontal dial test indicator from the BesTest series offers precise measurements with a maximum range of 0.0300 inch, a 0-15-0 dial reading, and a 0.001 inch graduation. It is designed for laboratory and industrial applications, adhering to DIN 2270 and EN ISO 9493 standards.
  • Horizontal indicator style
  • 1 inch dial diameter
  • Black dial color
  • Lever type with integrated shockproof system
  • Analogue display
  • Low measuring force
  • Rotating dial
  • Includes one contact point
  • 1/2 inch contact point length
  • 0.0800 inch ball diameter
  • 0.0300 inch range per revolution
  • Complies with DIN 2270 and EN ISO 9493 standards
  • Comes with a measurement report, wrench, mounting rod, declaration of conformity certificate, and a 0.08 inch insert