Learn More
MSC TESA Brown & Sharpe Horizontal Dial Test Indicator, 0.0300 in Max, 0.000500 in Accuracy
Catalog No. 5000528442
Encompass
This horizontal dial test indicator features a maximum measurement of 0.0300 in, a 0-15-0 dial reading, and an accuracy of 0.000500 in. It includes a 1 in diameter white dial with 0.000500 in graduation and meets DIN 2270 and EN ISO 9493 standards.
  • Lever type design.
  • Integrated shockproof system.
  • Analogue display.
  • Low measuring force.
  • Rotating dial.
  • Durable and reliable for long-term daily use.
  • Excellent value for a quality test indicator.
Catalog No. 50-005-28442 Supplier MSC Supplier No. 06433478
This horizontal dial test indicator features a maximum measurement of 0.0300 in, a 0-15-0 dial reading, and an accuracy of 0.000500 in. It includes a 1 in diameter white dial with 0.000500 in graduation and meets DIN 2270 and EN ISO 9493 standards.
  • Lever type design.
  • Integrated shockproof system.
  • Analogue display.
  • Low measuring force.
  • Rotating dial.
  • Durable and reliable for long-term daily use.
  • Excellent value for a quality test indicator.