Learn More
MSC TESA Brown & Sharpe Horizontal Dial Test Indicator, 0.0300 in Max, 0-15-0 Dial, 0.000500 in Graduation
Catalog No. 5000536686
Encompass
This horizontal dial test indicator provides precise measurements with a maximum range of 0.0300 inches and a dial graduation of 0.000500 inches. Designed for accuracy and durability, it features a shockproof system and a rotating dial for ease of use in various applications.
  • Lever type operation
  • Integrated shockproof system
  • Analogue display
  • Low measuring force
  • Rotating dial for flexible positioning
  • Dependable and sturdy construction
  • Easy to read black face and orange needle
Catalog No. 50-005-36686 Supplier MSC Supplier No. 09989757
This horizontal dial test indicator provides precise measurements with a maximum range of 0.0300 inches and a dial graduation of 0.000500 inches. Designed for accuracy and durability, it features a shockproof system and a rotating dial for ease of use in various applications.
  • Lever type operation
  • Integrated shockproof system
  • Analogue display
  • Low measuring force
  • Rotating dial for flexible positioning
  • Dependable and sturdy construction
  • Easy to read black face and orange needle