Learn More
MSC TESA Brown & Sharpe Horizontal Dial Test Indicator, 0.80 mm Max Measurement, 0-0.4-0 Dial Reading, 0.0100 mm Accuracy
Catalog No. 5000533391
Encompass
This horizontal dial test indicator offers precise measurements with a maximum range of 0.80 mm and an accuracy of 0.0100 mm. Its 0-0.4-0 dial reading and durable design, including an integrated shockproof system, make it a reliable tool for various inspection tasks.
  • Lever type
  • Integrated shockproof system
  • Analogue display
  • Rotating dial
  • Attachments included
  • One contact point
  • 2 mm insert
  • Measurement report
  • Wrench
  • Mounting rod
  • Plastic case
  • Declaration of conformity certificate
Catalog No. 50-005-33391 Supplier MSC Supplier No. 06226641
This horizontal dial test indicator offers precise measurements with a maximum range of 0.80 mm and an accuracy of 0.0100 mm. Its 0-0.4-0 dial reading and durable design, including an integrated shockproof system, make it a reliable tool for various inspection tasks.
  • Lever type
  • Integrated shockproof system
  • Analogue display
  • Rotating dial
  • Attachments included
  • One contact point
  • 2 mm insert
  • Measurement report
  • Wrench
  • Mounting rod
  • Plastic case
  • Declaration of conformity certificate