Learn More
MSC TESA Brown & Sharpe Micrometer Extension, 1.6-4.0 in Range, Intrimik Compatible
Catalog No. 5000533472
Encompass
This micrometer extension is designed for use with Intrimik Hole Micrometers, providing an expanded measurement range. It allows for precise measurements between 1.6 inches and 4.0 inches.
  • Functions as an extension accessory
  • Compatible with Intrimik hole micrometers
  • Enables measurements from 1.6 in
  • Extends measurement capability up to 4.0 in
  • Features an overall length of 6.0 in
  • Equipped with a satin chrome finish
Catalog No. 50-005-33472 Supplier MSC Supplier No. 06221469
This micrometer extension is designed for use with Intrimik Hole Micrometers, providing an expanded measurement range. It allows for precise measurements between 1.6 inches and 4.0 inches.
  • Functions as an extension accessory
  • Compatible with Intrimik hole micrometers
  • Enables measurements from 1.6 in
  • Extends measurement capability up to 4.0 in
  • Features an overall length of 6.0 in
  • Equipped with a satin chrome finish