Learn More
MSC TESA Brown & Sharpe Offset Ball Tip Probe, Tungsten Carbide, 3 mm Ball, 55 mm Length
Catalog No. 5000534619
Encompass
This TESA Brown & Sharpe offset ball tip probe is engineered for high-precision measurement tasks, particularly when used with height gages. Crafted from robust tungsten carbide, it offers a 3 mm ball diameter and a 55 mm probe length, ensuring reliable and accurate data collection in demanding metrology environments.
  • Offset design for improved accessibility in various measurement scenarios.
  • Constructed from tungsten carbide for superior durability and wear resistance.
  • Features a 3 mm ball diameter, ideal for precise contact and measurement.
  • Provides a 55 mm probe length for reaching confined spaces.
  • Equipped with a 6 mm fixation axis for secure mounting.
  • Ensures compatibility with Brown & Sharpe Micro-Hite height gages.
Catalog No. 50-005-34619 Supplier MSC Supplier No. 71547855
This TESA Brown & Sharpe offset ball tip probe is engineered for high-precision measurement tasks, particularly when used with height gages. Crafted from robust tungsten carbide, it offers a 3 mm ball diameter and a 55 mm probe length, ensuring reliable and accurate data collection in demanding metrology environments.
  • Offset design for improved accessibility in various measurement scenarios.
  • Constructed from tungsten carbide for superior durability and wear resistance.
  • Features a 3 mm ball diameter, ideal for precise contact and measurement.
  • Provides a 55 mm probe length for reaching confined spaces.
  • Equipped with a 6 mm fixation axis for secure mounting.
  • Ensures compatibility with Brown & Sharpe Micro-Hite height gages.