This steel rule is crafted from stainless steel for durability and corrosion resistance. It features a 5R Graduation style with measurements in 1/64, 1/100, and 1/32 inches, ensuring clear and precise readings. Its rigid design provides stability, while the silver, satin chrome-plated finish enhances the visibility of markings. With a 6-inch overall length and 3/4-inch width, it is a practical tool for various measuring tasks.
- Made from stainless steel, ensuring longevity and resistance to corrosion.
- Features 5R graduation style with measurements in 1/64, 1/100, and 1/32 inches, allowing for accurate readings.
- Rigid design ensures stability during use, providing consistent and reliable measurements.
- Silver, satin chrome-plated finish contributes to better visibility of markings.
- Practical tool for various measuring tasks.