Learn More
MSC TESA Brown & Sharpe Tapered Feeler Gage Set, 26 Piece, 0.0015-0.025 in
Catalog No. 5000530652
Encompass
This feeler gage set is a 26-piece collection of tapered gages, designed for precise measurements. Each leaf has a 3 in length and a 0.5 in width, and the set includes thicknesses ranging from 0.0015 to 0.025 in. The gages are constructed from durable steel.
  • Tapered leaf style
  • 26 leaves included
  • Thickness range of 0.0015 to 0.025 in
  • Leaf length of 3 in
  • Leaf width of 0.5 in
  • Constructed from steel
Catalog No. 50-005-30652 Supplier MSC Supplier No. 97232219
This feeler gage set is a 26-piece collection of tapered gages, designed for precise measurements. Each leaf has a 3 in length and a 0.5 in width, and the set includes thicknesses ranging from 0.0015 to 0.025 in. The gages are constructed from durable steel.
  • Tapered leaf style
  • 26 leaves included
  • Thickness range of 0.0015 to 0.025 in
  • Leaf length of 3 in
  • Leaf width of 0.5 in
  • Constructed from steel