Learn More
MSC TESA Brown & Sharpe Telescoping Gage Set, 5/16 to 6 in, 6 Pc, Satin Chrome
Catalog No. 5000529541
Encompass
This telescoping gage set is designed for measuring internal dimensions. It includes 6 pieces and features a satin chrome finish, with a measurement range from 5/16 inch to 6 inches. The set comes with a plastic case for convenient storage and protection.
  • Designed for measuring internal dimensions
  • Includes 6 pieces
  • Features a satin chrome finish
  • Comes with a plastic case for storage and protection
  • Measurement range from 5/16 inch to 6 inches
Catalog No. 50-005-29541 Supplier MSC Supplier No. 06457014
This telescoping gage set is designed for measuring internal dimensions. It includes 6 pieces and features a satin chrome finish, with a measurement range from 5/16 inch to 6 inches. The set comes with a plastic case for convenient storage and protection.
  • Designed for measuring internal dimensions
  • Includes 6 pieces
  • Features a satin chrome finish
  • Comes with a plastic case for storage and protection
  • Measurement range from 5/16 inch to 6 inches