Learn More
MSC TESA Brown & Sharpe Test Indicator Contact Point, 0.0400 in Ball Diameter, Carbide
Catalog No. 5000515164
Encompass
This contact point features a 0.0400 inch ball diameter and a carbide tip, designed for precise measurements. It is engineered for compatibility with specific dial test indicators, ensuring accurate and reliable performance in various applications.
  • Ball style
  • 0.0400 inch ball diameter
  • 12.70 mm contact point length
  • Carbide contact point material
Catalog No. 50-005-15164 Supplier MSC Supplier No. 97559652
This contact point features a 0.0400 inch ball diameter and a carbide tip, designed for precise measurements. It is engineered for compatibility with specific dial test indicators, ensuring accurate and reliable performance in various applications.
  • Ball style
  • 0.0400 inch ball diameter
  • 12.70 mm contact point length
  • Carbide contact point material