Learn More
MSC Tesa Tape Duct Tape, 48 mm, 550 m, 12.01 mil
Catalog No. 5000686132
Encompass
This duct tape features a polyethylene cloth backing and a natural rubber adhesive, engineered for secure and durable sealing applications.
  • Made from polyethylene cloth
  • Natural rubber adhesive
  • Tensile strength of 45.07 lb/in
  • Suitable for bundling, fastening, repair, seaming, and sealing
  • Silver color
Catalog No. 50-006-86132 Supplier MSC Supplier No. 41927690
This duct tape features a polyethylene cloth backing and a natural rubber adhesive, engineered for secure and durable sealing applications.
  • Made from polyethylene cloth
  • Natural rubber adhesive
  • Tensile strength of 45.07 lb/in
  • Suitable for bundling, fastening, repair, seaming, and sealing
  • Silver color