Learn More
MSC TESA Digital Display, Touch Screen, Multiple Inputs, 0.1 μm Resolution
Catalog No. 5000513522
Encompass
The TWIN-T20 digital display is a precision measurement instrument designed for simultaneous display of two measured values, suitable for use on the production floor and in test laboratories. It features multiple inputs for connecting probes and a wide range of measuring devices via integrated USB ports, ensuring accuracy with a quick and instinctive user display.
  • Large color touch screen with multiple visualizations.
  • Measures distance, diameter, and shape tolerances.
  • Supports static and dynamic measurements.
  • Multiple inputs: digital, inductive, and USB instruments.
  • Intuitive interface for setting tolerances.
  • Functions include calibration, data transfer, program storage.
  • Resolution: 0.1 μm / 0.00001 in.
  • Robust with IP65 front protection.
  • Battery-operated for portability.
Catalog No. 50-005-13522 Supplier MSC Supplier No. 17284019
The TWIN-T20 digital display is a precision measurement instrument designed for simultaneous display of two measured values, suitable for use on the production floor and in test laboratories. It features multiple inputs for connecting probes and a wide range of measuring devices via integrated USB ports, ensuring accuracy with a quick and instinctive user display.
  • Large color touch screen with multiple visualizations.
  • Measures distance, diameter, and shape tolerances.
  • Supports static and dynamic measurements.
  • Multiple inputs: digital, inductive, and USB instruments.
  • Intuitive interface for setting tolerances.
  • Functions include calibration, data transfer, program storage.
  • Resolution: 0.1 μm / 0.00001 in.
  • Robust with IP65 front protection.
  • Battery-operated for portability.