Learn More
Cayman Chemical Tesaglitazar, Purity: ≥98%, CAS: 251565-85-2, 50 mg
SDP
Catalog No. 502979604
Encompass_Preferred
A dual agonist of PPARα and γ (IC50s = 1 and 0.2 µM, respectively) that at 3 µM/kg/day for 3 weeks has been used to reduce insulin resistance in obese Zucker rats
Catalog No. 50-297-9604 Supplier Cayman Chemical Supplier No. 1679150 MG
A dual agonist of PPARα and γ (IC50s = 1 and 0.2 µM, respectively) that at 3 µM/kg/day for 3 weeks has been used to reduce insulin resistance in obese Zucker rats