Learn More
Medchemexpress LLC Tesirine intermediate-1 | 1430738-05-8 | 99.4% | C28H48N2O7Si2 | 250 MG
SDP
Catalog No. 5000357646
Encompass_Preferred
Tesirine intermediate-1 is an intermediate in the synthesis of Tesirine, a pyrrole benzodiazepine (PBD) dimer that acts as a DNA small channel crosslinker. This compound exhibits strong cytotoxicity and is utilized for synthesizing Antibody-Drug Conjugates (ADCs) whose payloads demonstrate potent anticancer cell activity.
  • Intermediate for advanced chemical synthesis
  • Functions as a pyrrole benzodiazepine (PBD) dimer
  • Acts as a DNA small channel crosslinker
  • Exhibits strong cytotoxicity
  • Key component for Antibody-Drug Conjugate (ADC) synthesis
  • Payloads demonstrate potent anticancer cell activity
Catalog No. 50-003-57646 Supplier Medchemexpress LLC Supplier No. 250MGHY47820
Tesirine intermediate-1 is an intermediate in the synthesis of Tesirine, a pyrrole benzodiazepine (PBD) dimer that acts as a DNA small channel crosslinker. This compound exhibits strong cytotoxicity and is utilized for synthesizing Antibody-Drug Conjugates (ADCs) whose payloads demonstrate potent anticancer cell activity.
  • Intermediate for advanced chemical synthesis
  • Functions as a pyrrole benzodiazepine (PBD) dimer
  • Acts as a DNA small channel crosslinker
  • Exhibits strong cytotoxicity
  • Key component for Antibody-Drug Conjugate (ADC) synthesis
  • Payloads demonstrate potent anticancer cell activity