Learn More
Carnabio Usa Inc TESK1/10UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 0910710
Encompass
TESK1, Full-length, Wild type, Amino Acid 1-626, NP_006276.2, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-1002