Learn More
Medchemexpress LLC Teslexivir hydrochloride | 1075281-70-7 | 98.28% | 679.04 | 25 MG
SDP
Catalog No. 5000347745
Encompass_Preferred
Teslexivir hydrochloride is a potent antiviral agent. It is a potent and selective inhibitor of the interaction between two essential viral proteins, E1 and E2. This association is a necessary step in the DNA replication and thus viral production for Human Papilloma Virus (HPV) 6 and 11. Teslexivir hydrochloride can be used for condyloma research.
  • Potent antiviral agent
  • Selective inhibitor of E1 and E2 viral protein interaction
  • Inhibits DNA replication and viral production for HPV 6 and 11
  • Suitable for condyloma research
Catalog No. 50-003-47745 Supplier Medchemexpress LLC Supplier No. 25MGHY109045A
Teslexivir hydrochloride is a potent antiviral agent. It is a potent and selective inhibitor of the interaction between two essential viral proteins, E1 and E2. This association is a necessary step in the DNA replication and thus viral production for Human Papilloma Virus (HPV) 6 and 11. Teslexivir hydrochloride can be used for condyloma research.
  • Potent antiviral agent
  • Selective inhibitor of E1 and E2 viral protein interaction
  • Inhibits DNA replication and viral production for HPV 6 and 11
  • Suitable for condyloma research