Learn More
Medchemexpress LLC Teslexivir hydrochloride | 1075281-70-7 | 98.3% | 679.04 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY109045A10MM/1ML
Encompass_Preferred
Teslexivir (BTA074) hydrochloride is a potent antiviral agent. It acts as a potent and selective inhibitor of the interaction between two essential viral proteins, E1 and E2, which is a necessary step in the DNA replication and viral production for Human Papilloma Virus (HPV) 6 and 11. Teslexivir hydrochloride is suitable for condyloma research.
  • Potent antiviral agent.
  • Selective inhibitor of the interaction between viral proteins E1 and E2.
  • Disrupts DNA replication and viral production for Human Papilloma Virus (HPV) 6 and 11.
  • Can be used for condyloma research.
Catalog No. 50-003-45666