Learn More
Medchemexpress LLC Teslexivir hydrochloride | 1075281-70-7 | 98.3% | 679.04 | 50 MG
SDP
Catalog No. 5000344459
Encompass_Preferred
Teslexivir (BTA074) hydrochloride is a potent antiviral agent. It acts as a potent and selective inhibitor of the interaction between two essential viral proteins, E1 and E2, which is a necessary step in the DNA replication and viral production for Human Papilloma Virus (HPV) 6 and 11. Teslexivir hydrochloride can be used for condyloma research.
  • Potent antiviral agent.
  • Potent and selective inhibitor of the interaction between E1 and E2 viral proteins.
  • Inhibits DNA replication and viral production for human papilloma virus (HPV) 6 and 11.
  • Can be used for condyloma research.
Catalog No. 50-003-44459 Supplier Medchemexpress LLC Supplier No. HY109045A100MG
Add to Cart
Add to Cart
Teslexivir (BTA074) hydrochloride is a potent antiviral agent. It acts as a potent and selective inhibitor of the interaction between two essential viral proteins, E1 and E2, which is a necessary step in the DNA replication and viral production for Human Papilloma Virus (HPV) 6 and 11. Teslexivir hydrochloride can be used for condyloma research.
  • Potent antiviral agent.
  • Potent and selective inhibitor of the interaction between E1 and E2 viral proteins.
  • Inhibits DNA replication and viral production for human papilloma virus (HPV) 6 and 11.
  • Can be used for condyloma research.