Learn More
Medchemexpress LLC Tesmilifene fumarate | 1185241-83-1 | 99.9% | 399.48 g/mol | C23H29NO5 | 10 MG
SDP
Catalog No. 5000213061
Encompass_Preferred
Tesmilifene fumarate is the fumarate salt of tesmilifene (DPPE fumarate), a small-molecule H1C receptor antagonist reported to potentiate a wide range of cytotoxic chemotherapies while offering some protection to normal cells. It is supplied for research use in solid and ready-to-use solution formats.
  • Potentiates cytotoxic chemotherapy in research settings.
  • Acts as an H1C receptor antagonist.
  • High purity (≈99.9%).
  • Molecular weight 399.48 g/mol; formula C23H29NO5.
  • Available in multiple vial sizes and as a 10 mM solution in DMSO.
  • Intended for research use and stored at -20°C.
Catalog No. 50-002-13061 Supplier Medchemexpress LLC Supplier No. HY10117910MG
Add to Cart
Add to Cart
Tesmilifene fumarate is the fumarate salt of tesmilifene (DPPE fumarate), a small-molecule H1C receptor antagonist reported to potentiate a wide range of cytotoxic chemotherapies while offering some protection to normal cells. It is supplied for research use in solid and ready-to-use solution formats.
  • Potentiates cytotoxic chemotherapy in research settings.
  • Acts as an H1C receptor antagonist.
  • High purity (≈99.9%).
  • Molecular weight 399.48 g/mol; formula C23H29NO5.
  • Available in multiple vial sizes and as a 10 mM solution in DMSO.
  • Intended for research use and stored at -20°C.