Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tesmilifene fumarate | 1185241-83-1 | MFCD09878272 | 99.9% | 399.48 | C23H29NO5 | 5 MG
SDP

Catalog No. 5000211237
Encompass_Preferred

Tesmilifene fumarate is a research-grade small molecule that acts as an H1C (histamine) receptor antagonist. It is reported to potentiate a wide range of cytotoxic agents and may offer some protection to normal cells. The compound is supplied for laboratory research and analytical applications.

  • Acts as an H1C receptor antagonist.
  • Potentiates a wide range of cytotoxic agents.
  • May offer some protection to normal cells.
  • Molecular formula C23H29NO5 and molecular weight 399.48.
  • Purity typically 99.85%.
  • Available in small research quantities, including 5 MG packs.

Catalog No. 50-002-11237 Supplier Medchemexpress LLC Supplier No. HY1011795MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Tesmilifene fumarate is a research-grade small molecule that acts as an H1C (histamine) receptor antagonist. It is reported to potentiate a wide range of cytotoxic agents and may offer some protection to normal cells. The compound is supplied for laboratory research and analytical applications.

  • Acts as an H1C receptor antagonist.
  • Potentiates a wide range of cytotoxic agents.
  • May offer some protection to normal cells.
  • Molecular formula C23H29NO5 and molecular weight 399.48.
  • Purity typically 99.85%.
  • Available in small research quantities, including 5 MG packs.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.