Learn More
Medchemexpress LLC Tesmilifene fumarate | 1185241-83-1 | MFCD09878272 | 99.9% | 399.48 g·mol⁻¹ | C23H29NO5 | 1 ML
SDP
Catalog No. 5000212126
Encompass_Preferred
Tesmilifene fumarate is the fumarate salt of tesmilifene, an H1C receptor antagonist and chemical sensitizer used in oncology research. Supplied as a 10 mM stock solution in DMSO (1 mL), it is intended for in vitro research applications including chemosensitization and cytotoxicity studies.
  • Ready-to-use 10 mM stock solution in DMSO
  • Concentrated 1 mL volume suitable for multiple assays
  • High purity (99.9%) for research use
  • Applicable to chemosensitization and cytotoxicity assays
  • Provided with molecular weight and CAS identifiers for reporting
Catalog No. 50-002-12126 Supplier Medchemexpress LLC Supplier No. HY10117910MM 1ML
Tesmilifene fumarate is the fumarate salt of tesmilifene, an H1C receptor antagonist and chemical sensitizer used in oncology research. Supplied as a 10 mM stock solution in DMSO (1 mL), it is intended for in vitro research applications including chemosensitization and cytotoxicity studies.
  • Ready-to-use 10 mM stock solution in DMSO
  • Concentrated 1 mL volume suitable for multiple assays
  • High purity (99.9%) for research use
  • Applicable to chemosensitization and cytotoxicity assays
  • Provided with molecular weight and CAS identifiers for reporting