Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tesmilifene fumarate | 1185241-83-1 | MFCD09878272 | 99.9% | 399.48 g/mol | C23H29NO5 | 25 MG
SDP

Catalog No. 5000212214
Encompass_Preferred

Tesmilifene fumarate (DPPE fumarate) is an H1C receptor antagonist used as a research reagent to potentiate a broad range of cytotoxic agents and may offer some protection to normal cells. It is supplied as a white to off-white solid and as a DMSO solution for laboratory use.

  • Potentiates a wide range of cytotoxic agents.
  • May provide partial protection to normal cells during cytotoxic exposure.
  • High reported purity (99.85%).
  • Molecular weight 399.48 g/mol; formula C23H29NO5.
  • Available as solid packs and as a 10 mM solution in DMSO.
  • Recommended storage: powder at -20 °C or 4 °C; in solvent at -80 °C or -20 °C.

Catalog No. 50-002-12214 Supplier Medchemexpress LLC Supplier No. HY10117925MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Tesmilifene fumarate (DPPE fumarate) is an H1C receptor antagonist used as a research reagent to potentiate a broad range of cytotoxic agents and may offer some protection to normal cells. It is supplied as a white to off-white solid and as a DMSO solution for laboratory use.

  • Potentiates a wide range of cytotoxic agents.
  • May provide partial protection to normal cells during cytotoxic exposure.
  • High reported purity (99.85%).
  • Molecular weight 399.48 g/mol; formula C23H29NO5.
  • Available as solid packs and as a 10 mM solution in DMSO.
  • Recommended storage: powder at -20 °C or 4 °C; in solvent at -80 °C or -20 °C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.