Learn More
Cayman Chemical Tesofensine, Purity: ≥95%, CAS: 195875-84-4, 100 mg
SDP
Catalog No. 502910905
Encompass_Preferred
A triple monoamine reuptake inhibitor; reduces body weight and food intake in a rat model of high-fat diet-induced obesity at 2 mg/kg
Catalog No. 50-291-0905 Supplier Cayman Chemical Supplier No. 35224100 MG
A triple monoamine reuptake inhibitor; reduces body weight and food intake in a rat model of high-fat diet-induced obesity at 2 mg/kg